Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

059637-500MG 059637-1G 059637-5G
101918-706EA 373.85 USD InStock
101918-706 101918-708 101918-710
1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one ≥95%
1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one
Hazard: Irritant


Formula: C₁₀H₁₁Cl₂N₅O
Melting Pt: 246 °C
MDL Number: MFCD04117980
CAS Number: 75438-54-9
Order Now
Specification Test Results
Melting point 246°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty