Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(hydroxymethyl)benzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

059812-500MG 059812-1G
101919-156EA 692.31 USD InStock
101919-156 101919-158
2-Fluoro-5-(hydroxymethyl)benzoic acid ≥95%
2-Fluoro-5-(hydroxymethyl)benzoic acid
Hazard: Irritant


Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 204…207 °C
MDL Number: MFCD01862001
CAS Number: 481075-38-1
Order Now
Specification Test Results
Melting point 204-207°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty