Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propan-2-yl-4-iodo-1H-pyrrole-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

059822-500MG 059822-1G 059822-5G
101919-204EA 803.08 USD InStock
101919-204 101919-206 101919-208
Propan-2-yl-4-iodo-1H-pyrrole-2-carboxylate ≥95%
Propan-2-yl-4-iodo-1H-pyrrole-2-carboxylate
Hazard: Irritant


Formula: C₈H₁₀INO₂
Melting Pt: 63…64 °C
MDL Number: MFCD18904322
CAS Number: 1296172-31-0
Order Now
Specification Test Results
Melting point 63-64°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty