Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorobenzoyl)piperidine hydrochloride

Supplier: MATRIX SCIENTIFIC MS

005950-5G 005950-25G
101789-832EA 36.92 USD InStock
101789-832 101789-834
4-(4-Fluorobenzoyl)piperidine hydrochloride
4-(4-Fluorobenzoyl)piperidine hydrochloride
Hazard: Irritant/Moisture Sensitive


Formula: C₁₂H₁₅ClFNO
MW: 243.71 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD00044912
CAS Number: 25519-78-2
Order Now
Specification Test Results
Melting point 222-224°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty