Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorophenyl)pyrazole

Supplier: MATRIX SCIENTIFIC MS

006107-5G 006107-25G
101790-092EA 67.69 USD InStock
101790-092 101790-094
3-(4-Fluorophenyl)pyrazole
3-(4-Fluorophenyl)pyrazole
Hazard: Irritant


Formula: C₉H₇FN₂
Melting Pt: 100…102 °C
MDL Number: MFCD00665857
CAS Number: 154258-82-9
Order Now
Specification Test Results
Melting point 100-102°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty