Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl methanesulfonylacetate ≥97%

Supplier: MATRIX SCIENTIFIC MS

006291-5G 006291-25G
101790-336EA 29.23 USD InStock
101790-336 101790-338
Methyl methanesulfonylacetate ≥97%
Methyl methanesulfonylacetate
Hazard: Irritant


Formula: C₄H₈O₄S
Melting Pt: 62…63 °C
MDL Number: MFCD00051842
CAS Number: 62020-09-1
Order Now
Specification Test Results
Melting point 62-63°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty