Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Iodomethyl)-4-(1H,1H-perfluorononyl)tetrahydrofuran

Supplier: MATRIX SCIENTIFIC MS

006333-250MG 006333-1G
101790-390EA 124.62 USD InStock
101790-390 101790-392
3-(Iodomethyl)-4-(1H,1H-perfluorononyl)tetrahydrofuran
3-(Iodomethyl)-4-(1H,1H-perfluorononyl)tetrahydrofuran
Hazard: Irritant


Formula: C₁₄H₁₀F₁₇IO
Melting Pt: 82…83 °C
MDL Number: MFCD01320758
CAS Number: 131771-31-8
Order Now
Specification Test Results
Melting point 82-83°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty