Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

006730-5G 006730-25G
101790-982EA 21.54 USD InStock
101790-982 101790-984
4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥97%
4,7-Dibromo-3-hydroxy-2-naphthoic acid
TCA listed: T
Hazard: Irritant


Formula: C₁₁H₆Br₂O₃
MW: 345.98 g/mol
Melting Pt: 251…253 °C
MDL Number: MFCD00046367
CAS Number: 1779-10-8
Order Now
Specification Test Results
Melting point 251-253°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty