Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylsulfonyl-1-(pyridin-2-yl)ethanone

Supplier: MATRIX SCIENTIFIC MS

006819-1G 006819-5G
101791-214EA 67.69 USD InStock
101791-214 101791-216
2-Methylsulfonyl-1-(pyridin-2-yl)ethanone
2-Methylsulfonyl-1-(pyridin-2-yl)ethanone
Hazard: Irritant


Formula: C₈H₉NO₃S
Melting Pt: 105…107 °C
MDL Number: MFCD01941305
CAS Number: 27302-93-8
Order Now
Specification Test Results
Melting point 105-107°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty