Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,4-dichloropyrimidine-6-carboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

006837-5G 006837-25G
101791-282EA 52.31 USD InStock
101791-282 101791-284
Methyl-2,4-dichloropyrimidine-6-carboxylate ≥97%
Methyl-2,4-dichloropyrimidine-6-carboxylate
Hazard: Irritant/Avoid Skin Contact


Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD01941317
CAS Number: 6299-85-0
Order Now
Specification Test Results
Melting point 54-56°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty