Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylsulphonyl)-2-nitrobenzoic acid

Supplier: MATRIX SCIENTIFIC MS

006881-5G 006881-25G 006881-100G
101791-446EA 21.54 USD InStock
101791-446 101791-448 102550-890
4-(Methylsulphonyl)-2-nitrobenzoic acid
4-(Methylsulphonyl)-2-nitrobenzoic acid
TCA listed: T
Hazard: Irritant


Formula: C₈H₇NO₆S
Melting Pt: 211-212°C
MDL Number: MFCD01941299
CAS Number: 110964-79-9
Order Now
Specification Test Results
Melting point 211-212°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty