Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)piperidine hydrochloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

007207-1G 007207-5G 007207-25G
101792-186EA 26.15 USD InStock
101792-186 101792-188 101792-190
4-(Trifluoromethyl)piperidine hydrochloride ≥98%
4-(Trifluoromethyl)piperidine hydrochloride
Hazard: Irritant/Irritant-Harmful


Formula: C₆H₁₁ClF₃N
Melting Pt: 155…156 °C
MDL Number: MFCD04971993
CAS Number: 155849-49-3
Order Now
Specification Test Results
Melting point 155-156°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty