Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzotriazole-4-sulfonic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

007234-5G 007234-25G
101792-218EA 1090.77 USD InStock
101792-218 101792-220
Benzotriazole-4-sulfonic acid ≥98%
Benzotriazole-4-sulfonic acid
Hazard: Irritant


Formula: C₆H₅N₃O₃S
Melting Pt: 300 °C (dec.)
MDL Number: MFCD03452776
CAS Number: 26725-50-8
Order Now
Specification Test Results
Melting point 300°C(dec)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty