Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylphenylhydrazine hydrochloride

Supplier: MATRIX SCIENTIFIC MS

007270-1G 007270-5G
101792-300EA 46.15 USD InStock
101792-300 101792-302
4-Ethylphenylhydrazine hydrochloride
4-Ethylphenylhydrazine hydrochloride
Hazard: Irritant/Irritant-Harmful


Formula: C₈H₁₃ClN₂
Melting Pt: 128-130°C
MDL Number: MFCD01861267
CAS Number: 53661-18-0
Order Now
Specification Test Results
Melting point 128-130°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty