Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyridin-3-yl)acetic acid hydrochloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

012513-5G 012513-25G
101802-724EA 15.38 USD InStock
101802-724 101802-726
2-(Pyridin-3-yl)acetic acid hydrochloride ≥98%
2-(Pyridin-3-yl)acetic acid hydrochloride
Hazard: Irritant


Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 161…163 °C
MDL Number: MFCD00012819
CAS Number: 6419-36-9
Order Now
Specification Test Results
Melting point 161-163°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty