Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)phenylglyoxal hydrate ≥97%

Supplier: MATRIX SCIENTIFIC MS

016081-250MG 016081-1G 016081-5G
101804-044EA 56.92 USD InStock
101804-044 101804-046 101804-048
4-(Trifluoromethyl)phenylglyoxal hydrate ≥97%
4-(Trifluoromethyl)phenylglyoxal hydrate
Hazard: Irritant


Formula: C₉H₅F₃O₂·xH₂O
Melting Pt: 87…89 °C
MDL Number: MFCD05664098
CAS Number: 1049746-22-6
Order Now
Specification Test Results
Melting point 87-89°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty