Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-2-pyrrolidin-1ylpyridine ≥97%

Supplier: MATRIX SCIENTIFIC MS

026305-1G 026305-5G
101822-048EA 189.23 USD InStock
101822-048 101822-050
5-Iodo-2-pyrrolidin-1ylpyridine ≥97%
5-Iodo-2-pyrrolidin-1ylpyridine
Hazard: Irritant


Formula: C₉H₁₁IN₂
MW: 274.11 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD07781186
CAS Number: 494771-62-9
Order Now
Specification Test Results
Melting point 106-108°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty