Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethyl-2-hydroxypyrimidine ≥98%

Supplier: MATRIX SCIENTIFIC MS

031255-25G 031255-100G
101831-326EA 27.69 USD InStock
101831-326 101831-328
4,6-Dimethyl-2-hydroxypyrimidine ≥98%
4,6-Dimethyl-2-hydroxypyrimidine
Hazard: Irritant


Formula: C₆H₈N₂O
Melting Pt: 202…204 °C
MDL Number: MFCD00006072
CAS Number: 108-79-2
Order Now
Specification Test Results
Melting point 202-204°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty