Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-2-chloropyridine ≥97%

Supplier: MATRIX SCIENTIFIC MS

035665-250MG 035665-1G
101839-606EA 26.15 USD InStock
101839-606 101839-608
4-Acetyl-2-chloropyridine ≥97%
4-Acetyl-2-chloropyridine
Hazard: Irritant-Harmful


Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 37…39 °C
MDL Number: MFCD07699406
CAS Number: 23794-15-2
Order Now
Specification Test Results
Melting point 37-39°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty