Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042510-500MG 042510-1G 042510-5G
101856-108EA 50.77 USD InStock
101856-108 101856-110 101856-112
Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant


Formula: C₁₂H₁₄N₂O₂
Melting Pt: 79-82°C
MDL Number: MFCD00139510
CAS Number: 81448-48-8
Order Now
Specification Test Results
Melting point 79-82°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty