Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

042529-500MG 042529-1G 042529-5G
101856-208EA 921.54 USD InStock
101856-208 101856-210 101856-212
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Hazard: Irritant


Formula: C₁₂H₈ClF₃N₂O₃
Melting Pt: 177…178 °C
MDL Number: MFCD00243689
CAS Number: 282523-49-3
Order Now
Specification Test Results
Melting point 177-178°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty