Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Nitrophenyl)pyrrolidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

042535-500MG 042535-1G 042535-5G
101856-238EA 181.54 USD InStock
101856-238 101856-240 101856-242
1-(4-Nitrophenyl)pyrrolidine ≥95%
1-(4-Nitrophenyl)pyrrolidine
Hazard: Irritant


Formula: C₁₀H₁₂N₂O₂
Melting Pt: 167…169 °C
MDL Number: MFCD00020819
CAS Number: 10220-22-1
Order Now
Specification Test Results
Melting point 167-169°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty