Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-{[(2-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

042545-500MG 042545-1G 042545-5G
101856-282EA 181.54 USD InStock
101856-282 101856-284 101856-286
2-{[(2-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%
2-{[(2-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid
Hazard: Irritant


Formula: C₁₄H₁₂ClNO₄S
Melting Pt: 96…98 °C
MDL Number: MFCD00172803
CAS Number: 250714-42-2
Order Now
Specification Test Results
Melting point 96-98°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty