Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Fluorobenzenesulfonyl)phenylalanine ≥95%

Supplier: MATRIX SCIENTIFIC MS

042563-500MG 042563-1G 042563-5G
101856-376EA 181.54 USD InStock
101856-376 101856-378 101856-380
N-(4-Fluorobenzenesulfonyl)phenylalanine ≥95%
N-(4-Fluorobenzenesulfonyl)phenylalanine
Hazard: Irritant


Formula: C₁₅H₁₄FNO₄S
Melting Pt: 108…110 °C
MDL Number: MFCD00173011
CAS Number: 40279-96-7
Order Now
Specification Test Results
Melting point 108-110°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty