Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Furylcarbonyl)-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

042586-500MG 042586-1G 042586-5G
101856-478EA 115.38 USD InStock
101856-478 101856-480 101856-482
1-(2-Furylcarbonyl)-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%
1-(2-Furylcarbonyl)-4-hydroxy-2-pyrrolidinecarboxylic acid
Hazard: Irritant


Formula: C₁₀H₁₁NO₅
Melting Pt: 224…227 °C
MDL Number: MFCD00202703
CAS Number: 251310-45-9
Order Now
Specification Test Results
Melting point 224-227°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty