Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Acetylphenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

042637-500MG 042637-1G 042637-5G
101856-732EA 747.69 USD InStock
101856-732 101856-734 101856-736
1-(4-Acetylphenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
1-(4-Acetylphenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile
Hazard: Irritant


Formula: C₁₃H₉N₃O₃
Melting Pt: 270…272 °C
MDL Number: MFCD00172915
CAS Number: 446276-09-1
Order Now
Specification Test Results
Melting point 270-272°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty