Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042656-500MG 042656-1G
101856-836EA 278.46 USD InStock
101856-836 101856-838
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant


Formula: C₁₁H₈ClF₃N₂O₂
MW: 292.65 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD01833013
CAS Number: 353258-31-8
Order Now
Specification Test Results
Melting point 132-134°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty