Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042686-500MG 042686-1G 042686-5G
101857-002EA 181.54 USD InStock
101857-002 101857-004 101857-006
Methyl-2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate ≥95%
Methyl-2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate
Hazard: Irritant


Formula: C₁₂H₁₀ClNO₂S
Melting Pt: 138…140 °C
MDL Number: MFCD00792936
CAS Number: 337924-65-9
Order Now
Specification Test Results
Melting point 138-140°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty