Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

042728-500MG 042728-1G
101857-218EA 421.54 USD InStock
101857-218 101857-220
1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic acid ≥95%
1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic acid
Hazard: Irritant


Formula: C₁₀H₉N₃O₂
Melting Pt: 213…215 °C
MDL Number: MFCD02187274
CAS Number: 477847-81-7
Order Now
Specification Test Results
Melting point 213-215°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty