Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-(2,5-dimethoxyphenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042748-500MG 042748-1G 042748-5G
101857-320EA 181.54 USD InStock
101857-320 101857-322 101857-324
Ethyl-4-(2,5-dimethoxyphenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(2,5-dimethoxyphenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Hazard: Irritant


Formula: C₁₆H₂₀N₂O₅
Melting Pt: 218…219 °C
MDL Number: MFCD00397696
CAS Number: 292855-70-0
Order Now
Specification Test Results
Melting point 218-219°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty