Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(1,3-Benzodioxol-5-yl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

042754-500MG 042754-1G
101857-356EA 375.38 USD InStock
101857-356 101857-358
6-(1,3-Benzodioxol-5-yl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%
6-(1,3-Benzodioxol-5-yl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
Hazard: Irritant


Formula: C₁₂H₁₀N₂O₅
Melting Pt: 243 °C (dec.)
MDL Number: MFCD03787253
CAS Number: 860610-20-4
Order Now
Specification Test Results
Melting point 243°C(dec)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty