Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1,6-dimethyl-4-phenyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042760-500MG 042760-1G 042760-5G
101857-388EA 181.54 USD InStock
101857-388 101857-390 101857-392
Ethyl-1,6-dimethyl-4-phenyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-1,6-dimethyl-4-phenyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Hazard: Irritant


Formula: C₁₅H₁₈N₂O₂S
Melting Pt: 150…152 °C
MDL Number: MFCD01112452
CAS Number: 108958-81-2
Order Now
Specification Test Results
Melting point 150-152°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty