Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3,5-bis(allyloxy)benzenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042784-500MG 042784-1G 042784-5G
101857-514EA 181.54 USD InStock
101857-514 101857-516 101857-518
Methyl-3,5-bis(allyloxy)benzenecarboxylate ≥95%
Methyl-3,5-bis(allyloxy)benzenecarboxylate
Hazard: Irritant


Formula: C₁₄H₁₆O₄
Melting Pt: 30…33 °C
MDL Number: MFCD04117781
CAS Number: 135710-38-2
Order Now
Specification Test Results
Melting point 30-33°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty