Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(3-(3-(hydroxymethyl)-8-methoxychroman-4-yl)-3-methylthioureido)acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042981-500MG 042981-5G
101858-562EA 1855.38 USD InStock
101858-562 101858-566
Ethyl-2-(3-(3-(hydroxymethyl)-8-methoxychroman-4-yl)-3-methylthioureido)acetate ≥95%
Ethyl-2-(3-(3-(hydroxymethyl)-8-methoxychroman-4-yl)-3-methylthioureido)acetate
Hazard: Irritant


Formula: C₁₇H₂₄N₂O₅S
MW: 368.46 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00243893
CAS Number: 303148-56-3
Order Now
Specification Test Results
Melting point 151-153°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty