Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trifluoro-N-(7-oxo-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-yl)acetamide ≥95%

Supplier: MATRIX SCIENTIFIC MS

043066-500MG 043066-5G
101858-974EA 556.92 USD InStock
101858-974 101858-978
2,2,2-Trifluoro-N-(7-oxo-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-yl)acetamide ≥95%
2,2,2-Trifluoro-N-(7-oxo-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-yl)acetamide
Hazard: Irritant


Formula: C₁₂H₈F₃NO₄
Melting Pt: 226…220 °C
MDL Number: MFCD03102675
CAS Number: 138621-69-9
Order Now
Specification Test Results
Melting point 226-220°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty