Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-2-bromo-2-(5-nitro-2-pyridinyl)malonate ≥90%

Supplier: MATRIX SCIENTIFIC MS

043075-500MG 043075-5G
101859-026EA 278.46 USD InStock
101859-026 101859-030
Dimethyl-2-bromo-2-(5-nitro-2-pyridinyl)malonate ≥90%
Dimethyl-2-bromo-2-(5-nitro-2-pyridinyl)malonate
Hazard: Irritant


Formula: C₁₀H₉BrN₂O₆
Melting Pt: 84…86 °C
MDL Number: MFCD03787057
CAS Number: 860650-18-6
Order Now
Specification Test Results
Melting point 84-86°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty