Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043199-500MG 043199-1G 043199-5G
101859-636EA 375.38 USD InStock
101859-636 101859-638 101859-640
Methyl-5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate ≥95%
Methyl-5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate
Hazard: Irritant


Formula: C₁₄H₁₂F₃N₃O₄
Melting Pt: 46…48 °C
MDL Number: MFCD00138717
CAS Number: 303995-97-3
Order Now
Specification Test Results
Melting point 46-48°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty