Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(2-cyanoanilino)acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043440-500MG 043440-1G 043440-5G
101860-804EA 278.46 USD InStock
101860-804 101860-806 101860-808
Ethyl-2-(2-cyanoanilino)acetate ≥95%
Ethyl-2-(2-cyanoanilino)acetate
Hazard: Irritant


Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 95…96 °C
MDL Number: MFCD00141059
CAS Number: 87223-76-5
Order Now
Specification Test Results
Melting point 95-96°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty