Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

043447-500MG 043447-1G 043447-5G
101860-840EA 747.69 USD InStock
101860-840 101860-842 101860-844
4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid ≥95%
4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
Hazard: Irritant


Formula: C₁₂H₈F₃NO₂S
MW: 287.26 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD00142049
CAS Number: 144059-85-8
Order Now
Specification Test Results
Melting point 174-176°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty