Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-(3,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043478-500MG 043478-1G 043478-5G
101861-004EA 375.38 USD InStock
101861-004 101861-006 101861-008
Ethyl-1-(3,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate ≥95%
Ethyl-1-(3,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate
Hazard: Irritant


Formula: C₁₂H₈Cl₅N₃O₂
Melting Pt: 110…113 °C
MDL Number: MFCD04117789
CAS Number: 854019-67-3
Order Now
Specification Test Results
Melting point 110-113°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty