Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2,4-difluorophenol ≥95%

Supplier: MATRIX SCIENTIFIC MS

043566-500MG 043566-1G 043566-5G
101861-472EA 84.62 USD InStock
101861-472 101861-474 101861-476
5-Amino-2,4-difluorophenol ≥95%
5-Amino-2,4-difluorophenol
Hazard: Irritant


Formula: C₆H₅F₂NO
MW: 145.11 g/mol
Melting Pt: 189…190 °C
MDL Number: MFCD04973718
CAS Number: 113512-71-3
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 189-190°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty