Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

043614-500MG 043614-1G 043614-5G
101861-710EA 323.08 USD InStock
101861-710 101861-712 101861-714
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid ≥95%
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid
Hazard: Irritant


Formula: C₁₀H₇ClN₂O₂
MW: 222.63 g/mol
Melting Pt: 210…213 °C
MDL Number: MFCD03425733
CAS Number: 313674-12-3
Order Now
Specification Test Results
Melting point 210-213°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty