Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone ≥95%

Supplier: MATRIX SCIENTIFIC MS

043668-500MG 043668-1G 043668-5G
101861-976EA 181.54 USD InStock
101861-976 101861-978 101861-980
1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone ≥95%
1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone
Hazard: Irritant


Formula: C₁₁H₁₃ClO₃
Melting Pt: 73…74 °C
MDL Number: MFCD04126369
CAS Number: 172739-45-6
Order Now
Specification Test Results
Melting point 73-74°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty