Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043695-500MG 043695-1G 043695-5G
101862-104EA 100 USD InStock
101862-104 101862-106 101862-108
Ethyl-5-oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylate ≥95%
Ethyl-5-oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylate
Hazard: Irritant


Formula: C₉H₁₀N₂O₃S
Melting Pt: 175…177 °C
MDL Number: MFCD00085067
CAS Number: 32084-53-0
Order Now
Specification Test Results
Melting point 175-177°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty