Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl 2,3-dichlorobenzenesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043722-500MG 043722-1G 043722-5G
101862-234EA 375.38 USD InStock
101862-234 101862-236 101862-238
Pentafluorophenyl 2,3-dichlorobenzenesulfonate ≥95%
Pentafluorophenyl 2,3-dichlorobenzenesulphonate
Hazard: Irritant


Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 117-119°C
MDL Number: MFCD06659539
CAS Number: 885949-54-2
Order Now
Specification Test Results
Melting point 117-119°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty