Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl 4-cyanobenzenesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043725-500MG 043725-1G 043725-5G
101862-252EA 375.38 USD InStock
101862-252 101862-254 101862-256
Pentafluorophenyl 4-cyanobenzenesulfonate ≥95%
Pentafluorophenyl 4-cyanobenzenesulphonate
Hazard: Irritant


Formula: C₁₃H₄F₅NO₃S
MDL Number: MFCD06659542
CAS Number: 885949-56-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty