Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chloro-2-[2-(4-chloro-2-nitrophenyl)hydrazono]acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043761-500MG 043761-1G 043761-5G
101862-440EA 181.54 USD InStock
101862-440 101862-442 101862-444
Ethyl-2-chloro-2-[2-(4-chloro-2-nitrophenyl)hydrazono]acetate ≥95%
Ethyl-2-chloro-2-[2-(4-chloro-2-nitrophenyl)hydrazono]acetate
Hazard: Irritant


Formula: C₁₀H₉Cl₂N₃O₄
MW: 306.11 g/mol
Melting Pt: 104…105 °C
MDL Number: MFCD06200942
CAS Number: 112091-27-7
Order Now
Specification Test Results
Melting point 104-105°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty