Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(6-Chloropyridazin-3-yl)-1H-indole ≥95%

Supplier: MATRIX SCIENTIFIC MS

043784-500MG 043784-1G 043784-5G
101862-542EA 52.31 USD InStock
101862-542 101862-544 101862-546
3-(6-Chloropyridazin-3-yl)-1H-indole ≥95%
3-(6-Chloropyridazin-3-yl)-1H-indole
Hazard: Irritant


Formula: C12H8ClN3
Melting Pt: 270…272 °C
CAS Number: 129287-26-9
Order Now
Specification Test Results
Melting point 270-272°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty