Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-(1H-imidazol-1-yl)nicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043791-500MG 043791-1G 043791-5G
101862-578EA 181.54 USD InStock
101862-578 101862-580 101862-582
Methyl-6-(1H-imidazol-1-yl)nicotinate ≥95%
Methyl-6-(1H-imidazol-1-yl)nicotinate
Hazard: Irritant


Formula: C₁₀H₉N₃O₂
Melting Pt: 185…187 °C
MDL Number: MFCD00140820
CAS Number: 111205-01-7
Order Now
Specification Test Results
Melting point 185-187°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty