Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol ≥95%

Supplier: MATRIX SCIENTIFIC MS

043858-500MG 043858-1G 043858-5G
101862-940EA 1296.92 USD InStock
101862-940 101862-942 101862-944
[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol ≥95%
[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol
Hazard: Irritant


Formula: C₈H₇F₃N₂OS
Melting Pt: 88…92 °C
MDL Number: MFCD01936006
CAS Number: 338982-22-2
Order Now
Specification Test Results
Melting point 88-92°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty